CHEMets® Ammonia Visual Test Kit (HBA)
Catalog # 77674-718
Supplier: CHEMetrics
New Product
CAS Number:
Specifications
- Chemical method:Hydroxybenzyl Alcohol (HBA)
- Application:Drinking water;Clean surface water;Waste water;Storm water
Specifications
About this item
This ammonia test uses the hydroxybenzyl alcohol method of analysis.K-1420 is a perfect fit for environmental, wastewater, stormwater, aquaculture, or any application where a range of ammonia concentrations may be measured and rapid results required. This kit can be used to determine free ammonia concentration in the presence of monochloramine.
- Range: 0 to 4 and 0 to 80 ppm MDL: 0.125 ppm Method: Hydroxybenzyl Alcohol (HBA)
Delivery: The K-1420 ammonia CHEMets® test kit (HBA) comes in a plastic case and contains everything needed to perform 30 tests (except distilled water): CHEMets®refill, flat comparator, stabilizer solution, catalyzer solution, activator solution, 25 ml sample cup, 3 ml syringe, and instructions.