$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
TRC Carisoprodol (1.0 mg/ml in Methanol)
TRC Carisoprodol (1.0 mg/ml in Methanol)
77987-735
:
LGC Standards
New Product
:
TRC Carisoprodol (1.0 mg/ml in Methanol)
77987-735
:
LGC Standards
:
TRC-KIT2020-5X1ML
New Product
:
Matrix:
Methanol
CAS No.:
78-44-4
Description:
Carisoprodol (1.0 mg/ml in Methanol)
Size:
5×1 ml