$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
TRC cis-Montelukast
TRC cis-Montelukast
77989-751
:
LGC Standards
New Product
:
TRC cis-Montelukast
77989-751
:
LGC Standards
:
TRC-M568005-5MG
New Product
:
CAS No.:
774538-96-4
Description:
cis-Montelukast
Size:
5 mg