ContactUsLinkComponent
Silica Comparator Flat 1 to 10 ppm
  77674-684
New Product
 :  
undefined
Silica Comparator Flat 1 to 10 ppm
  77674-684
 :  C-9010
New Product
 :  
  • Chemical method:
    Heteropoly Blue
C-9010 High Range Silica Comparator is a component of K-9010 Silica CHEMets Visual Test Kit and is designed to be compared against the color formed in R-9010 Silica CHEMets Refill. The comparator's range is applicable to many industrial applications. Color standards are blue in color and cover a range of 1 to 10 ppm SiO₂