$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
TRC Vanillic Acid
TRC Vanillic Acid
77995-171
:
LGC Standards
New Product
:
TRC Vanillic Acid
77995-171
:
LGC Standards
:
TRC-V097350-100MG
New Product
:
CAS No.:
121-34-6
Description:
Vanillic Acid
Size:
100 mg