TRC N-Ethyl-N-nitroso-2-propanamine-d5 (1.0 mg/ml in Methanol)
Catalog # 77987-758
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Matrix:Methanol
- Description:N-Ethyl-N-nitroso-2-propanamine-d5 (1.0 mg/ml in Methanol)
- Size:1×1ml