Lipomed 6-Acetylmorphine-D3.HCl.trihydrate, 1 mg/ml in Methanol (as free base)
Catalog # 77976-195
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Matrix:Methanol
- Description:6-Acetylmorphine-D3.HCl.trihydrate, 1 mg/ml in Methanol (as free base)
- Size:1 ml