Orders LinkContactUsLinkComponent
Flat Chlorine Comparator
DPD (free and total), 0 - 5.0 ppm, 0, 1.0, 1.5, 2.0, 2.5, 3.0, 3.5, 4.0, 5.0 ppm
  77674-598
 :   CHEMetrics
 :  C-2506
Flat Chlorine Comparator
  77674-598
 :  
Flat Chlorine Comparator
Flat Chlorine Comparator
DPD (free and total), 0 - 5.0 ppm, 0, 1.0, 1.5, 2.0, 2.5, 3.0, 3.5, 4.0, 5.0 ppm
 :  
$29.10
Each
C-2506 High Range Chlorine Comparator is a component of K-2504 Chlorine (free and total) CHEMets Visual Test Kit and is designed to be compared against the color formed in R-2500 Chlorine CHEMets Refill. The comparator's range is applicable for many disinfection and drinking water applications. Color standards are pink in color and cover a range of 0 to 5.0 ppm chlorine (Cl₂).
  • Chemical method:
    DPD (free and total)
  • Increments:
    0, 1.0, 1.5, 2.0, 2.5, 3.0, 3.5, 4.0, 5.0 ppm
  • Range:
    0 - 5.0 ppm