TRC Clonidine Hydrochloride
New Product
- CAS No.:4205-91-8
Frequently Bought Together
CrossLab Accessories for Thermal Conductivity Detector (TCD), Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Tubing and Fittings for 8453/8454 UV Systems
: AGILENT TECHNOLOGIES, INC (CSD)
O-Rings and Fittings for ICP-OES
: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Ultimate Union for GC, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Injectors for Fully Demountable Torches, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Reciprocating Dissolution Screens, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
Capillaries for HPLC, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Fittings for HPLC, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
Pump Valves and Supplies for HPLC, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Dissolution Full Flow Filters, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
FRITS REPLACEMENT TWIST 10UM/15UM PK20
: GLEN RESEARCH CORP MS
pH Gradient Buffers
: Thermo Scientific
AVS 6/7 Supplies for ICP-OES, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Tubing for HPLC, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
Tubing Kits for ICP-OES
: AGILENT TECHNOLOGIES, INC (CSD)
Cleaning and Maintenance Supplies for GC/MS, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)