Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
TRC N-Nitroso Phenylephrine (1 mg/ml in methanol)
TRC N-Nitroso Phenylephrine (1 mg/ml in methanol)
TRC N-Nitroso Phenylephrine (1 mg/ml in methanol)
Catalog # 77987-743
Supplier:
LGC Standards
New Product
CAS Number:
Expand image
TRC N-Nitroso Phenylephrine (1 mg/ml in methanol)
Catalog # 77987-743
Supplier:
LGC Standards
Supplier Number:
TRC-KIT3492-1X1ML
New Product
CAS Number:
Specifications
Matrix:
Methanol
CAS No.:
78658-64-7
Description:
N-Nitroso Phenylephrine (1 mg/ml in methanol)
Size:
1×1ml
Specifications
About this item
Product Details
Product Details