Paragon Scientific Sulfur @ 0.075% (750 ppm) in heavy mineral oil
Catalog # 77975-762
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Matrix:Heavy Mineral Oil
- CAS No.:7704-34-9
- Description:Sulfur @ 0.075% (750 ppm) in heavy mineral oil
- Size:100 ml
- Certification:ISO 17034
- Storage Conditions:RT
- Shipping Conditions:RT
- Molecular Weight:32.065