Paragon Scientific Relative Density 25 °C (Nominal: 0.8352 @ 25 °C)
77975-959
New Product
:
- Pack type:Glass bottle
- Description:Relative density 25 °C (Nominal: 0.8352 @ 25 °C), Standard type: Single component
- Size:500 ml
- Shipping conditions: Room temperature