$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
TRC Pyriproxyfen-D4
TRC Pyriproxyfen-D4
77992-465
:
LGC Standards
New Product
:
TRC Pyriproxyfen-D4
77992-465
:
LGC Standards
:
TRC-P998852-1MG
New Product
:
CAS No.:
2446366-95-4
Description:
Pyriproxyfen-D4
Size:
1 mg