CHEMets® Persulfate Visual Test Kit
Product Details & Documents
- Range: 0 to 5.6 and 7 to 70 ppm as Na₂S₂O₈ MDL: 0.35 ppm Method: Ferric Thiocyanate
Delivery information: Kit comes in a plastic case and contains everything needed to perform 30 tests:refill, low and high range comparators, 25 ml sample cup, and instructions.
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Chemical method:Ferric thiocyanate
- Application:Industrial cooling water;Municipal water treatment systems;Food and beverages