$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
TRC (4-Sulfamoylphenyl)hydrazine Hydrochloride
TRC (4-Sulfamoylphenyl)hydrazine Hydrochloride
Catalog # 77993-626
Supplier:
LGC Standards
New Product
CAS Number:
Expand image
TRC (4-Sulfamoylphenyl)hydrazine Hydrochloride
Catalog # 77993-626
Supplier:
LGC Standards
Supplier Number:
TRC-S699078-1G
New Product
CAS Number:
Specifications
CAS No.:
17852-52-7
Description:
(4-Sulfamoylphenyl)hydrazine Hydrochloride
Size:
1 g
Specifications
About this item
Product Details
Product Details