$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
TRC Monocaprylin
TRC Monocaprylin
TRC Monocaprylin
77989-742
:
LGC Standards
New Product
:
TRC Monocaprylin
77989-742
:
LGC Standards
:
TRC-M567253-1G
New Product
:
CAS No.:
502-54-5
Description:
Monocaprylin
Size:
1 g