ContactUsLinkComponent
Hardness Titration Test Kit 100 to 1000 ppm
  77674-778
New Product
 :  
undefined
Hardness Titration Test Kit 100 to 1000 ppm
  77674-778
 :  K-4585
New Product
 :  
  • Chemical method:
    EDTA (total)
  • Application:
    Drinking water;Surface water;Saline water
Features of titrets technology: Dependable, sealed single-use titrants Fast, easy procedure, no need to count drops No burettes or other equipment necessary Perform a titration ANYWHERE
  • Range: 100 to 1000 ppm as CaCO₃ MDL: 100 ppm Method: EDTA (total) Increments: 100, 110, 120, 130, 140, 150, 160, 180, 200, 250, 300, 350, 400, 500, 700, 1000 ppm CaCO₃
 : The K-4585 Total Hardness Titrets® test kit (100 to 1000 ppm) comes in a cardboard box and contains everything needed to perform 30 tests: thirty Titrets® ampoules with valve assemblies, plastic titrettor, 25 ml sample cup, and instructions.