Order Entry
Puerto Rico
ContactUsLinkComponent
Manganese Instrumental Test 0 to 30 ppm
Manganese Instrumental Test 0 to 30 ppm
Catalog # 77674-814
Supplier:  CHEMetrics
New Product
CAS Number:  
undefined
Manganese Instrumental Test 0 to 30 ppm
Catalog # 77674-814
Supplier:  CHEMetrics
Supplier Number:  K-6503
New Product
CAS Number:  

Specifications

  • Chemical method:
    Periodate
  • Application:
    Drinking water;Natural water;Treated waste water effluent

Specifications

About this item

  • Range: 0 to 30.0 ppm Method: Periodate
Delivery: Kit comes in a cardboard box and contains everything needed to perform 30 tests: thirty ampoules, activator solution, 25 ml sample cup, 1.0 ml syringe, ampoule blank, and instructions. This kit requires the use of a CHEMetrics direct-readout photometer or a spectrophotometer capable of accepting a 13 mm diameter round cell. Instrument sold separately.