$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
TRC 1-Bromobutane-2,2-d2
TRC 1-Bromobutane-2,2-d2
Catalog # 77978-965
Supplier:
LGC Standards
New Product
CAS Number:
Expand image
TRC 1-Bromobutane-2,2-d2
Catalog # 77978-965
Supplier:
LGC Standards
Supplier Number:
TRC-B681834-100MG
New Product
CAS Number:
Specifications
CAS No.:
55724-40-8
Description:
1-Bromobutane-2,2-d2
Size:
100 mg
Specifications
About this item
Product Details
Product Details