Paragon Scientific Type PSU3600, Viscosity Reference Standard
Catalog # 77975-935
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Description:Type PSU3600, viscosity reference standard
- Size:500 ml
- Shipping Conditions:RT
- Storage Temperature:RT
- Certification:ISO 17034
Specifications
About this item
- Standard type: Single component