Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
TRC Isopilocarpine Hydrochloride
TRC Isopilocarpine Hydrochloride
TRC Isopilocarpine Hydrochloride
77987-403
:
LGC Standards
New Product
:
TRC Isopilocarpine Hydrochloride
77987-403
:
LGC Standards
:
TRC-I821970-25MG
New Product
:
CAS No.:
28958-85-2
Description:
Isopilocarpine Hydrochloride
Size:
25 mg