$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
TRC 4’-Hydroxy Tamoxifen (contains up to 10% E isomer)
TRC 4’-Hydroxy Tamoxifen (contains up to 10% E isomer)
77986-847
:
LGC Standards
New Product
:
TRC 4’-Hydroxy Tamoxifen (contains up to 10% E isomer)
77986-847
:
LGC Standards
:
TRC-H954730-2.5MG
New Product
:
CAS No.:
82413-23-8
Description:
4’-Hydroxy Tamoxifen (contains up to 10% E isomer)
Size:
2.5 mg