ContactUsLinkComponent
Iron In Brine Comparator Flat 100 to 1000 ppm
Catalog # 77674-646
Supplier:  CHEMetrics
New Product
CAS Number:  
undefined
Iron In Brine Comparator Flat 100 to 1000 ppm
Catalog # 77674-646
Supplier:  CHEMetrics
Supplier Number:  C-6012
New Product
CAS Number:  
Specifications
  • Chemical method:
    Ferric Thiocyanate (Iron in brine)
  • Cat. No.:
    77674-646
  • Increments:
    100, 200, 300, 400, 500, 600, 700, 800, 1000 mg/L
  • Range:
    100 - 1000 mg/L
Specifications
About this item
C-6012 High Range Iron in Brine Comparator is a component of K-6002 Iron in Brine CHEMets Visual Test Kit and is designed to be compared against the color formed in R-6002 Iron in Brine CHEMets Refill. The comparator's range is applicable to many industrial applications. Color standards are orange in color and cover a range of 100 to 1000 mg/L.