TRC Tetraethyleneglycol Bisibuprofen Ester
New Product
- CAS No.:1161014-75-0
Frequently Bought Together
Vaplock™ Chemical Exhaust Filters and Color Indicators, Cole-Parmer
: Antyila Scientific
CrossLab Spray Chambers for Atomic Absorption, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
ZORBAX Ion Exchange, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
Empty Cartridges and Empty Cartridges with Pre-installed Frits, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Tubing for HPLC, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Purged Packed Inlet for GC, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
pH Gradient Buffers
: Thermo Scientific
Upchurch Scientific® PFA and PFA HP Fluoropolymer Tubing, IDEX Health & Science
: Upchurch Scientific
Splitless Liners for GC
: AGILENT TECHNOLOGIES, INC (CSD)
Pump Valves and Supplies for HPLC, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab GC Fittings, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
Capillaries for HPLC, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
Upchurch Scientific® Precolumn Filters, IDEX Health & Science
: Upchurch Scientific
CrossLab Reciprocating Dissolution Screens, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Dissolution Full Flow Filters, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
FRITS REPLACEMENT TWIST 10UM/15UM PK20
: GLEN RESEARCH CORP MS
Upchurch Scientific® Manifolds, Multi-Port Connectors, IDEX Health & Science
: Upchurch Scientific
CrossLab Torches for 5100/5110 and 5800/5900 ICP-OES, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)