0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Standards
Organic Standards
Lipomed Phenobarbital, 1 mg/ml in Methanol
Lipomed Phenobarbital, 1 mg/ml in Methanol
Lipomed Phenobarbital, 1 mg/ml in Methanol
Catalog # 77976-231
Supplier:
LGC Standards
New Product
CAS Number:
Expand image
Lipomed Phenobarbital, 1 mg/ml in Methanol
Catalog # 77976-231
Supplier:
LGC Standards
Supplier Number:
LPM-PHB-376-FA-1LM
New Product
CAS Number:
Specifications
Matrix:
Methanol
CAS No.:
50-06-6
Description:
Phenobarbital, 1 mg/ml in Methanol
Size:
1 ml
Specifications
About this item
Product Details
Product Details