Order Entry
Puerto Rico
ContactUsLinkComponent
Sulfite In White Wine Titrets® Test Kit (10 to 100 ppm)
Sulfite In White Wine Titrets® Test Kit (10 to 100 ppm)
Catalog # 77674-880
Supplier:  CHEMetrics
New Product
CAS Number:  
undefined
Sulfite In White Wine Titrets® Test Kit (10 to 100 ppm)
Catalog # 77674-880
Supplier:  CHEMetrics
Supplier Number:  K-9610W
New Product
CAS Number:  

Specifications

  • Chemical method:
    Ripper (Sulfite in wine)
  • Application:
    Food and beverages

Specifications

About this item

Features of titrets titration test kit technology: Dependable, sealed single-use titrants Fast, easy procedure, no need to count drops No burettes or other equipment necessary Perform a titration ANYWHERE

  • Range: 10 to 100 ppm as SO₂ MDL: 10 ppm Method: Ripper (Sulfite in Wine) Increments: 10, 11, 12, 13, 14, 15, 16, 18, 20, 25, 30, 35, 40, 50, 70, 100 ppm
Delivery: Sulfite in white wine testing kit comes ready to use. Each set comes in a cardboard box and contains everything needed to perform 10 SO₂ tests: ten ampoules, ten valve assemblies, and instructions. Results for this SO₂ test are acceptable for white wines (although they can have an error up to 10 ppm). This titration test kit is not recommended for use with red wines or white wines containing ascorbic acid or tannin. These wines often give false high test results.