Order Entry
Puerto Rico
ContactUsLinkComponent
CHEMets® Ammonia Visual HR Test Kit (Nessler 1000 to 10000 ppm)
CHEMets® Ammonia Visual HR Test Kit (Nessler 1000 to 10000 ppm)
Catalog # 77674-728
Supplier:  CHEMetrics
New Product
CAS Number:  
undefined
CHEMets® Ammonia Visual HR Test Kit (Nessler 1000 to 10000 ppm)
Catalog # 77674-728
Supplier:  CHEMetrics
Supplier Number:  K-1520C
New Product
CAS Number:  

Specifications

  • Chemical method:
    Direct Nesslerization
  • Application:
    Drinking water;Clean surface water;Waste water;Storm water

Specifications

About this item

Delivery: The K-1520C ammonia HR CHEMets® test kit (1000 to 10,000 ppm) comes in a plastic case and contains everything needed to perform 30 tests (except distilled water): 30 HR CHEMets® ampoules, a color comparator, fixed volume pipet, 25 ml sample cup, and instructions. This kit is applicable for wastewater, environmental, aquaculture or any application where high concentrations of ammonia may be found in water.
Caution: Contains mercury. Dispose according to local, state or federal laws.