$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
TRC Apatinib-d8 Hydrochloride
TRC Apatinib-d8 Hydrochloride
77977-657
:
LGC Standards
New Product
:
TRC Apatinib-d8 Hydrochloride
77977-657
:
LGC Standards
:
TRC-A726152-5MG
New Product
:
CAS No.:
2468771-44-8
Description:
Apatinib-d8 Hydrochloride
Size:
5 mg