Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
TRC 1-(4-(Benzyloxy)-3-nitrophenyl)-2-bromoethanone
TRC 1-(4-(Benzyloxy)-3-nitrophenyl)-2-bromoethanone
TRC 1-(4-(Benzyloxy)-3-nitrophenyl)-2-bromoethanone
77978-382
:
LGC Standards
New Product
:
TRC 1-(4-(Benzyloxy)-3-nitrophenyl)-2-bromoethanone
77978-382
:
LGC Standards
:
TRC-B287165-1G
New Product
:
CAS No.:
43229-01-2
Description:
1-(4-(Benzyloxy)-3-nitrophenyl)-2-bromoethanone
Size:
1 g