ContactUsLinkComponent
Persulfate Comparator Flat 7 to 70 ppm
Persulfate Comparator Flat 7 to 70 ppm
Catalog # 77674-670
Supplier:  CHEMetrics
New Product
CAS Number:  
undefined
Persulfate Comparator Flat 7 to 70 ppm
Catalog # 77674-670
Supplier:  CHEMetrics
Supplier Number:  C-7870
New Product
CAS Number:  
Specifications
  • Chemical method:
    Ferric Thiocyanate
Specifications
About this item
C-7870 High Range Persulfate Comparator is a component of K-7870 Persulfate CHEMets Visual Test Kit and is designed to be compared against the color formed in R-5510 Peroxide/Persulfate CHEMets Refill. The comparator's range is applicable to many industrial applications. Color standards are orange in color and cover a range of 7 to 70 ppm sodium persulfate.