$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
TRC 3-Dechloro Sertraline Hydrochloride
TRC 3-Dechloro Sertraline Hydrochloride
TRC 3-Dechloro Sertraline Hydrochloride
Catalog # 77981-210
Supplier:
LGC Standards
New Product
CAS Number:
Expand image
TRC 3-Dechloro Sertraline Hydrochloride
Catalog # 77981-210
Supplier:
LGC Standards
Supplier Number:
TRC-D226650-10MG
New Product
CAS Number:
Specifications
CAS No.:
79646-00-7
Description:
3-Dechloro Sertraline Hydrochloride
Size:
10 mg
Specifications
About this item
Product Details
Product Details