Order Entry
Puerto Rico
ContactUsLinkComponent
 
Paragon Scientific 15W40 Viscosity Check Oil (Kinematic Viscosity, CCS Dynamic Viscosity and Density)
New Product
 
undefined
Paragon Scientific 15W40 Viscosity Check Oil (Kinematic Viscosity, CCS Dynamic Viscosity and Density)
New Product

 

  • Shipping Conditions:
    RT
  • Storage Temperature:
    RT