Lipomed Morphine-D3.monohydrate, 1 mg/ml in Methanol (as free base)
Catalog # 77976-190
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Matrix:Methanol
- CAS No.:202656-19-7
- Description:Morphine-D3.monohydrate, 1 mg/ml in Methanol (as free base)
- Size:1 ml