$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
TRC D-Phenylalanine-d5
TRC D-Phenylalanine-d5
77991-586
:
LGC Standards
New Product
:
TRC D-Phenylalanine-d5
77991-586
:
LGC Standards
:
TRC-P319412-25MG
New Product
:
CAS No.:
362049-55-6
Description:
D-Phenylalanine-d5
Size:
25 mg