Paragon Scientific Type C10, Flow Cup Viscosity
Catalog # 77975-514
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Description:Type C10, flow cup viscosity
- Size:500 ml
- Shipping Conditions:RT
- Storage Temperature:RT
- Certification:ISO 17034
Specifications
About this item
- Standard type: Single component