Orders LinkContactUsLinkComponent
Hydrogen Peroxide Activator Solution Pack
Hydrogen Peroxide Activator Solution Pack
DPD
Catalog # 77674-558
Supplier:   CHEMetrics
Supplier #:  A-5500
Hydrogen Peroxide Activator Solution Pack
Catalog # 77674-558
Supplier:  CHEMetrics
CAS Number:  
Hydrogen Peroxide Activator Solution Pack
Hydrogen Peroxide Activator Solution Pack
DPD
CAS Number:  
Price
$17.28
Log in for availability
Pack of 6 (10ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The A-5500 accessory pack supplies enough solution to perform at least 200 tests. Qty. per Pack: Six 10 ml bottles.
Specifications
Specifications
  • Chemical method:
    DPD