$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
TRC Thifensulfuron
TRC Thifensulfuron
Catalog # 77993-983
Supplier:
LGC Standards
New Product
CAS Number:
Expand image
TRC Thifensulfuron
Catalog # 77993-983
Supplier:
LGC Standards
Supplier Number:
TRC-T176770-100MG
New Product
CAS Number:
Specifications
CAS No.:
79277-67-1
Description:
Thifensulfuron
Size:
100 mg
Specifications
About this item
Product Details
Product Details