ContactUsLinkComponent
Hydrazine Comparator Round 0 to 0.5 ppm
Hydrazine Comparator Round 0 to 0.5 ppm
Catalog # 77674-628
Supplier:  CHEMetrics
New Product
CAS Number:  
undefined
Hydrazine Comparator Round 0 to 0.5 ppm
Catalog # 77674-628
Supplier:  CHEMetrics
Supplier Number:  C-5005
New Product
CAS Number:  
Specifications
  • Chemical method:
    PDMAB
  • Cat. No.:
    77674-628
  • Increments:
    0, 0.01, 0.03, 0.05, 0.07, 0.1, 0.3, 0.5 ppm
  • Range:
    0 - 0.5 ppm
Specifications
About this item
C-5005 Hydrazine Comparator is a component of K-5005 Hydrazine CHEMets Visual Test Kit and is designed to be compared against the color formed in R-5005 Hydrazine CHEMets Refill. The comparator's range is applicable for many boiler water applications. Color standards are yellow in color and cover a range of 0 to 0.5 ppm hydrazine.