$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
TRC 4,4'-Difluorobenzhydryl Bromide
TRC 4,4'-Difluorobenzhydryl Bromide
77978-742
:
LGC Standards
New Product
:
TRC 4,4'-Difluorobenzhydryl Bromide
77978-742
:
LGC Standards
:
TRC-B519105-100MG
New Product
:
CAS No.:
345-90-4
Description:
4,4'-Difluorobenzhydryl Bromide
Size:
100 mg