Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
TRC 2-(Bismethyl)aminomethylcyclohexanone Hydrochloride
TRC 2-(Bismethyl)aminomethylcyclohexanone Hydrochloride
77978-835
:
LGC Standards
New Product
:
TRC 2-(Bismethyl)aminomethylcyclohexanone Hydrochloride
77978-835
:
LGC Standards
:
TRC-B585445-2.5G
New Product
:
CAS No.:
42036-65-7
Description:
2-(Bismethyl)aminomethylcyclohexanone Hydrochloride
Size:
2.5 g