$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
TRC (R)-2-((4-Aminophenethyl)amino)-1-phenylethanol Hydrochloride
TRC (R)-2-((4-Aminophenethyl)amino)-1-phenylethanol Hydrochloride
77977-331
:
LGC Standards
New Product
:
TRC (R)-2-((4-Aminophenethyl)amino)-1-phenylethanol Hydrochloride
77977-331
:
LGC Standards
:
TRC-A622675-1G
New Product
:
CAS No.:
521284-22-0
Description:
(R)-2-((4-Aminophenethyl)amino)-1-phenylethanol Hydrochloride
Size:
1 g