Order Entry
Puerto Rico
ContactUsLinkComponent
Silica ULR Comparator Round 0 to 0.2 ppm
Silica ULR Comparator Round 0 to 0.2 ppm
Catalog # 11027-208
Supplier:  CHEMetrics
New Product
CAS Number:  
undefined
Silica ULR Comparator Round 0 to 0.2 ppm
Catalog # 11027-208
Supplier:  CHEMetrics
Supplier Number:  C-9011
New Product
CAS Number:  

Specifications

  • Chemical method:
    Heteropoly Blue

Specifications

About this item

C-9011 Silica Comparator is a component of K-9011 Silica ULR CHEMets Visual Test Kit and is designed to be compared against the color formed in R-9011 Silica ULR CHEMets Refill. The comparator's range is applicable to many industrial applications. Color standards are blue in color and cover a range of 0 to 0.20 ppm SiO₂