TRC Prochlorperazine Dimaleate
New Product
- CAS No.:84-02-6
Frequently Bought Together
PEEK Tubing, Trajan Scientific and Medical
: Trajan Scientific and Medical
Upchurch Scientific® PEEK™ Tubing, IDEX Health & Science
: Upchurch Scientific
Vaplock™ Chemical Exhaust Filters and Color Indicators, Cole-Parmer
: Antyila Scientific
Captiva Syringe Filters, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Spray Chambers for Atomic Absorption, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
Stay Safe Caps & Accessories for HPLC, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
pH Gradient Buffers
: Thermo Scientific
CrossLab Fittings and Connectors for ICP-MS, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Pre-Optics Cone and Windows for ICP-OES, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
AVS 6/7 Supplies for ICP-OES, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
Capillaries for HPLC, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
Pump Valves and Supplies for HPLC, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Connector Tubes from Spray Chamber to Torch for ICP-MS, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
Agilent Prep HPLC Columns, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Instrument Supplies for GC, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
Disintegration Tubes, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)