ContactUsLinkComponent
Nitrate Acidifier Solution Pack
Nitrate Acidifier Solution Pack
Catalog # 77674-572
Supplier:  CHEMetrics
New Product
CAS Number:  
undefined
Nitrate Acidifier Solution Pack
Catalog # 77674-572
Supplier:  CHEMetrics
Supplier Number:  A-6901
New Product
CAS Number:  
Specifications
  • Chemical method:
    Zinc Reduction
Specifications
About this item
The accessory pack supplies enough solution to perform approximately 200 tests. Qty. per pack: Six 20 ml Bottles.