Order Entry
Puerto Rico
ContactUsLinkComponent
TRC (4-(((2-Amino-4-oxo-3,4-dihydropteridin-6-yl)methyl)(nitroso)amino)benzoyl)-L-glutamic Acid (1 mg/mL in DMSO)
TRC (4-(((2-Amino-4-oxo-3,4-dihydropteridin-6-yl)methyl)(nitroso)amino)benzoyl)-L-glutamic Acid (1 mg/mL in DMSO)
Catalog # 77987-744
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
TRC (4-(((2-Amino-4-oxo-3,4-dihydropteridin-6-yl)methyl)(nitroso)amino)benzoyl)-L-glutamic Acid (1 mg/mL in DMSO)
Catalog # 77987-744
Supplier:  LGC Standards
Supplier Number:  TRC-KIT3497-1X1ML
New Product
CAS Number:  

Specifications

  • Matrix:
    DMSO
  • CAS No.:
    26360-21-4
  • Description:
    (4-(((2-Amino-4-oxo-3,4-dihydropteridin-6-yl)methyl)(nitroso)amino)benzoyl)-L-glutamic Acid (1 mg/ml in DMSO)
  • Size:
    1×1 ml

Specifications

About this item