TRC O-Desmethyl Metoprolol
Supplier: LGC Standards
New Product
Certificates
About this item
Specifications
- CAS No.:62572-94-5
Specifications
Frequently Bought Together
Vaplock™ Chemical Exhaust Filters and Color Indicators, Cole-Parmer
Supplier: Antyila Scientific
Manual Crimpers and Decappers, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Tubing Kits for ICP-OES
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Sealing Mats for InfinityLab Well Plates, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Column Installation Pre-Swaging Tools for GC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
pH Gradient Buffers
Supplier: Thermo Scientific
AVS 6/7 Supplies for ICP-OES, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Captiva Syringe Filters, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Spray Chambers for Atomic Absorption, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Dissolution Baskets, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Upchurch Scientific® Flat-Bottom Fittings, For Tubing Ø = ¹/₁₆" (1.6 mm), IDEX Health & Science
Supplier: Upchurch Scientific
CrossLab Dissolution Full Flow Filters, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Accessories for Tubing for HPLC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
FRITS REPLACEMENT TWIST 10UM/15UM PK20
Supplier: GLEN RESEARCH CORP MS
CrossLab Ultra-Microcells for UV-Vis and UV-Vis-NIR Systems, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Liner O-Rings for GC
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Tools for GC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)