Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Home
Standards
Organic Standards
TRC 2 4'-Methylenebis(phenyl Isocyanate)
TRC 2 4'-Methylenebis(phenyl Isocyanate)
Catalog # 77988-704
Supplier:
LGC Standards
New Product
CAS Number:
Expand image
TRC 2 4'-Methylenebis(phenyl Isocyanate)
Catalog # 77988-704
Supplier:
LGC Standards
Supplier Number:
TRC-M261935-10MG
New Product
CAS Number:
Specifications
CAS No.:
5873-54-1
Description:
2 4'-Methylenebis(phenyl Isocyanate)
Size:
10 mg
Specifications
About this item
Product Details
Product Details