Order Entry
Puerto Rico
ContactUsLinkComponent
Lipomed Hydroxyzine.2HCl, 1 mg/ml in Methanol (as free base)
Lipomed Hydroxyzine.2HCl, 1 mg/ml in Methanol (as free base)
Catalog # 77976-182
Supplier:  LGC Standards
New Product
CAS Number:  
Lipomed Hydroxyzine.2HCl, 1 mg/ml in Methanol (as free base)
Catalog # 77976-182
Supplier:  LGC Standards
Supplier Number:  LPM-HDX-1733-HC-1LM
New Product
CAS Number:  

Specifications

  • Matrix:
    Methanol
  • CAS No.:
    2192-20-3
  • Description:
    Hydroxyzine.2HCl, 1 mg/ml in Methanol (as free base)
  • Size:
    1 ml
  • Certification:
    ISO 17034
  • Certification:
    ISO 17034

Specifications

About this item