Order Entry
Puerto Rico
ContactUsLinkComponent
Nitrate Instrumental Test 0 to 50 ppm
Nitrate Instrumental Test 0 to 50 ppm
Catalog # 77674-830
Supplier:  CHEMetrics
New Product
CAS Number:  
undefined
Nitrate Instrumental Test 0 to 50 ppm
Catalog # 77674-830
Supplier:  CHEMetrics
Supplier Number:  K-6933
New Product
CAS Number:  

Specifications

  • Chemical method:
    Cadmium reduction
  • Application:
    Industrial waste water influent and effluent;Industrial process water;Boiler water;Cooling water;Surface and ground water;Potable water

Specifications

About this item

  • Range: 0 to 50.0 ppm as NO₃ Method: Cadmium reduction
Delivery: Nitrate instrumental test kit comes in a cardboard box and contains everything needed to perform 30 tests (except distilled water): thirty ampoules, thirty cadmium foil packs, 25 ml sample cup, reaction tube with lid, 3.0 ml syringe, ampoule blank, and instructions. This kit requires the use of a CHEMetrics direct-readout photometer or a spectrophotometer capable of accepting a 13 mm diameter round cell. Instrument sold separately.