Order Entry
Puerto Rico
ContactUsLinkComponent
TRC 8-Bromo-7-(2-butyn-1-yl)-3,7-dihydro-3-methyl-1-[(4-methyl-2-quinazolinyl)methyl]-1H-purine-2,6-dione
TRC 8-Bromo-7-(2-butyn-1-yl)-3,7-dihydro-3-methyl-1-[(4-methyl-2-quinazolinyl)methyl]-1H-purine-2,6-dione
Catalog # 77978-959
Supplier:  LGC Standards
New Product
CAS Number:  
TRC 8-Bromo-7-(2-butyn-1-yl)-3,7-dihydro-3-methyl-1-[(4-methyl-2-quinazolinyl)methyl]-1H-purine-2,6-dione
Catalog # 77978-959
Supplier:  LGC Standards
Supplier Number:  TRC-B680510-1G
New Product
CAS Number:  

Specifications

  • CAS No.:
    853029-57-9
  • Description:
    8-Bromo-7-(2-butyn-1-yl)-3,7-dihydro-3-methyl-1-[(4-methyl-2-quinazolinyl)methyl]-1H-purine-2,6-dione
  • Size:
    1 g

Specifications

About this item